Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MultiTox-Fluor™ Multiplex Cytotoxicity Assay

Catalog No. PRG9200 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9200 10 mL
PRG9201 5 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRG9200 Supplier Promega Supplier No. G9200
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Assay buffer; bis-AAF-R110 Substrate; GF-AFC Substrate

The MultiTox-Fluor Multiplex Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay that measures the number of live and dead cells simultaneously in culture wells.

  • A Fluorescent Assay That Measures Both Cell Viability and Cytotoxicity
  • Assay normalizes data for cell number
  • Reduces false-positive and false-negative results

Order Info

Each 10mL aliquot is sufficient for 100 assays at 100μL/assay in a 96-well format or 400 assays at 25μL/assay in a 384-well format

Specifications

Content And Storage -30°C to -10°C
Detection Method Homogeneous, Fluorescence
For Use With (Equipment) Microplate Reader
Includes 1 x 10μL GF-AFC substrate, 1 x 10μL bis-AAF-R110 substrate, 1 x 10mL Assay Buffer
Quantity 10 mL
Sufficient For 100 Assays at 100μL/assay in a 96-well format or 400 assays at 25μL/assay in a 384-well format

Patent pending.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.