Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUM1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB290830 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290830 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290830 Supplier Novus Biologicals Supplier No. NBP178760F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MUM1 Polyclonal antibody specifically detects MUM1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen MUM1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EXPAND1, FLJ14868, FLJ22283, melanoma associated antigen (mutated) 1, melanoma ubiquitous mutated protein, MGC131891, MGC163315, MUM-1HSPC211, Mutated melanoma-associated antigen 1, Protein expandere, PWWP domain-containing protein MUM1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 125 and 175 of human Melanoma Associated Antigen (Mutated) 1 using the numbering given in entry NP_116242.2 (GeneID 84939).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Melanoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 84939
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.