Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUPP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP149975 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149975 Supplier Novus Biologicals Supplier No. NBP149975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MUPP1 Polyclonal specifically detects MUPP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen MUPP1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias DKFZp781P216, FLJ25909, FLJ90240, Multi-PDZ domain protein 1, multiple PDZ domain protein, MUPP1FLJ34626
Gene Symbols MPDZ
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1125 and 1150 of human Multiple PDZ Domain Protein using the numbering given in entry NP_003820.2 (GeneID 8777).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8777
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.