Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MuRF1/TRIM63 Antibody (M316), Novus Biologicals™
SDP

Catalog No. NB387831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387831 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387831 Supplier Novus Biologicals Supplier No. NBP323424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MuRF1/TRIM63 Monoclonal antibody specifically detects MuRF1/TRIM63 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MuRF1/TRIM63
Applications Western Blot
Classification Monoclonal
Clone M316
Conjugate Unconjugated
Dilution Western Blot 1:250
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63
Host Species Mouse
Immunogen Clone M316 was generated from a synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal half of human MuRF1/TRIM63. This peptide sequence is highly conserved in rat and mouse MuRF1/TRIM63, and has 50% homology to MuRF2 (TRIM-55). [UniProt# Q969Q1]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84676
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.