Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Murine anti-PON1, Clone: KRJ1, ZeptoMetrix

Murine Monoclonal Antibody
$2276.00 - $2388.00
Specifications
Antigen | PON1 |
---|---|
Clone | KRJ1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-158-012
![]() |
ZeptoMetrix
0801012 |
100 μg |
Each for $2,388.00
|
|
|||||
22-158-014
![]() |
ZeptoMetrix
0801095 |
1 mg |
Each for $2,276.00
|
|
|||||
Description
PON1 (arylesterase/paraoxonase) is a polymorphic enzyme closely associated with high-density lipoproteins (HDL). It is located on chromosome 7 along with 2 other paraoxanase genes, PON2 and PON3. It is a 355 amino acid, 43kDa glycoprotein produced in the liver and secreted into the bloodstream. Gene polymorphisms occur in both coding and non-coding regions. Two major polymorphisms in the coding region are L55M and Q192R. Q192R polymorphisms result in three major functional phenotypes Q/Q, Q/R and RR. PON1 hydrolyses organophosphates and prevents the oxidation of low-density lipoproteins (LDL). It may inhibit the progression of atherosclerosis by inhibition of LDL oxidation. Levels of PON1 in serum vary among individuals and are decreased in chronic renal failure, liver cirrhosis, diabetes, peripheral neuropathy, and familial hypercholesterolemia.Specifications
PON1 | |
Western Blot | |
Unconjugated | |
RUO | |
Partially purified PON1 from human plasma | |
Primary | |
Store at −10°C or below. Avoid freeze/thaw cycles. Thawed material may be stored at 4°C for short-term use. |
KRJ1 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title