Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP22671549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22671549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22671549 Supplier Novus Biologicals Supplier No. NBP221671JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Murine Hepatitis Virus A59 nsp9 Monoclonal specifically detects Murine Hepatitis Virus A59 nsp9 in Virus samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen nsp9
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2C6.H1
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Non-structural protein 9, p12
Gene Symbols nsp9
Host Species Mouse
Immunogen This Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1) was produced in mice by repeated immunizations with E.coli derived full-length Murine Hepatitis Virus A59 nsp9 protein. This protein is part of the viral replicase polyprotein. (Uniprot: P0C6V0)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1489749
Test Specificity No cross reactivity occurs with SARS CoV nsp9.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.