Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mus81 Antibody (mta30 2g10/3), FITC, Novus Biologicals™
SDP

Catalog No. NB1002064F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002064F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002064F Supplier Novus Biologicals Supplier No. NB1002064F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mus81 Monoclonal specifically detects Mus81 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Mus81
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mta30 2g10/3
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias crossover junction endonuclease MUS81, EC 3.1.22, EC 3.1.22.-, FLJ21012, FLJ44872, MUS81 endonuclease homolog (S. cerevisiae), MUS81 endonuclease homolog (yeast), SLX3, SLX3 structure-specific endonuclease subunit homolog
Gene Symbols MUS81
Host Species Mouse
Immunogen Human MUS81
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 80198
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.