Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Musashi-2 Rabbit anti-Human, Clone: RM422, Novus Biologicals™
SDP

Catalog No. NB119698 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119698 25 μg
NB119697 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB119698 Supplier Novus Biologicals Supplier No. NBP30917525UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Musashi-2 Monoclonal specifically detects Musashi-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Musashi-2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone RM422
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:400
Formulation PBS, 50% Glycerol, 1% BSA
Gene Alias FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2
Host Species Rabbit
Immunogen A peptide corresponding to the internal region of human Musashi-2
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 124540
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.