Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscarinic Acetylcholine Receptor M2/CHRM2 Antibody (585713), HRP, Novus Biologicals™
SDP

Catalog No. NB176327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176327 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176327 Supplier Novus Biologicals Supplier No. FAB6377H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen Muscarinic Acetylcholine Receptor M2/CHRM2
Applications Immunohistochemistry
Classification Monoclonal
Clone 585713
Conjugate HRP
Formulation PBS
Gene Alias 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CHRM2, Accession # P08172
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1129
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.