Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscarinic Acetylcholine Receptor M3/CHRM3 Antibody (580011), Biotin, Novus Biologicals™
SDP

Catalog No. NB376631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376631 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376631 Supplier Novus Biologicals Supplier No. FAB6378B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Muscarinic Acetylcholine Receptor M3/CHRM3 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M3/CHRM3 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Muscarinic Acetylcholine Receptor M3/CHRM3
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 580011
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias cholinergic receptor, muscarinic 3, HM3, m3 muscarinic receptor, muscarinic acetylcholine receptor M3
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CHRM3, Accession # P20309
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1131
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.