Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscarinic Acetylcholine Receptor M3/CHRM3 Antibody (SR2111), Novus Biologicals™
SDP

Catalog No. NB259709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259709 100 μg
NB260053 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259709 Supplier Novus Biologicals Supplier No. NBP321828100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Muscarinic Acetylcholine Receptor M3/CHRM3 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M3/CHRM3 in Human samples. It is validated for Western Blot

Specifications

Antigen Muscarinic Acetylcholine Receptor M3/CHRM3
Applications Western Blot
Classification Monoclonal
Clone SR2111
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias cholinergic receptor, muscarinic 3, HM3, m3 muscarinic receptor, muscarinic acetylcholine receptor M3
Host Species Rabbit
Immunogen A synthesized peptide derived from human Muscarinic Acetylcholine Receptor M3/CHRM3 (Uniprot #: P20309)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1131
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.