Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscarinic Acetylcholine Receptor M4/CHRM4 Antibody, Novus Biologicals™
SDP

Catalog No. NB093895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB093895 Supplier Novus Biologicals Supplier No. NBP303052

Rabbit Polyclonal Antibody

Muscarinic Acetylcholine Receptor M4/CHRM4 Polyclonal specifically detects Muscarinic Acetylcholine Receptor M4/CHRM4 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Muscarinic Acetylcholine Receptor M4/CHRM4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias cholinergic receptor, muscarinic 4, HM4, M4R, muscarinic acetylcholine receptor M4
Host Species Rabbit
Immunogen A synthetic peptide of human Muscarinic Acetylcholine Receptor M4/CHRM4.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1132
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.