Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscarinic Acetylcholine Receptor M5/CHRM5 Antibody (590123) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB401883 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401883 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401883 Supplier Novus Biologicals Supplier No. FAB10323J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Muscarinic Acetylcholine Receptor M5/CHRM5
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 590123
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias cholinergic receptor, muscarinic 5, HM5, MGC41838, muscarinic acetylcholine receptor M5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CHRM5, Met1-Pro532, Accession # P08912
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1133
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.