Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody, HRP, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Muscle Phosphofructokinase/PFKM/PFK-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS with No Preservative |
| Gene Alias | 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase |
| Gene Symbols | PFKM |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Phosphofructokinase, Muscle using the numbering given in entry NP_000280.1 (GeneID 5213). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.