Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscleblind-like 1 Antibody (1H1), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB360387 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360387 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360387 Supplier Novus Biologicals Supplier No. NBP250062MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Muscleblind-like 1 Monoclonal antibody specifically detects Muscleblind-like 1 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Muscleblind-like 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1H1
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp686P06174, EXP40, EXP42, EXPKIAA0428EXP35, MBNL, muscleblind (Drosophila)-like, muscleblind-like (Drosophila), muscleblind-like protein 1, Triplet-expansion RNA-binding protein
Host Species Mouse
Immunogen Full-length recombinant human muscleblind-like 1 protein expressed in and purified from E. coli. [UniProt# Q9NR56]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 4154
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.