Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVK Antibody (OTI 1D7), PE, Novus Biologicals™
SDP

Catalog No. NB287762 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287762 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287762 Supplier Novus Biologicals Supplier No. NBP272837PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MVK Monoclonal antibody specifically detects MVK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MVK
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone OTI 1D7
Conjugate PE
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK
Host Species Mouse
Immunogen Full length human recombinant protein of human MVK (NP_000422) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4598
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.