Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MVK Antibody (SR1713), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322554100UL
Description
MVK Monoclonal antibody specifically detects MVK in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
MVK | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
SR1713 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK | |
A synthesized peptide derived from human MVK (Uniprot #: Q03426) | |
100 μg | |
metabolism | |
4598 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction