Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVK Antibody (SR1713), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$309.50 - $423.00

Specifications

Antigen MVK
Clone SR1713
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50
Applications Western Blot, Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB259270
SDP
View Documents
Novus Biologicals
NBP322554100UL
100 μg
Each for $423.00
Only null left
Add to Cart
 
NB259286
SDP
View Documents
Novus Biologicals
NBP32255450UL
50 μg
Each for $309.50
Only null left
Add to Cart
 
Description

Description

MVK Monoclonal antibody specifically detects MVK in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications

Specifications

MVK
Western Blot 1:500-1:2000, Flow Cytometry 1:50
Monoclonal
Purified
RUO
Human
EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK
A synthesized peptide derived from human MVK (Uniprot #: Q03426)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1713
Western Blot, Flow Cytometry
Unconjugated
Rabbit
metabolism
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
4598
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.