Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVP Antibody (VP2897R), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB390543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390543 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390543 Supplier Novus Biologicals Supplier No. NBP308801JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MVP Monoclonal antibody specifically detects MVP in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MVP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VP2897R
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias LRPVAULT1, Lung resistance-related protein, major vault protein
Host Species Rabbit
Immunogen Recombinant full-length human MVP protein (Uniprot: Q14764)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9961
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.