Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MxA/Mx1 Antibody (683), Novus Biologicals™
SDP

Catalog No. NBP243786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243786 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243786 Supplier Novus Biologicals Supplier No. NBP243786

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

MxA/Mx1 Monoclonal specifically detects MxA/Mx1 in Human samples. It is validated for Western Blot.

Specifications

Antigen MxA/Mx1
Applications Western Blot
Classification Monoclonal
Clone 683
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS with No Preservative
Gene Alias human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse)
Gene Symbols MX1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human MX1. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 4599
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.