Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MxA/Mx1 Antibody (OTI2G12), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP272838A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272838A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272838A94 Supplier Novus Biologicals Supplier No. NBP272838AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MxA/Mx1 Monoclonal specifically detects MxA/Mx1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MxA/Mx1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2G12
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse)
Gene Symbols MX1
Host Species Mouse
Immunogen Full length recombinant protein of human MxA/Mx1 (NP_002453) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4599
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.