Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mxi1 Rabbit anti-Human, DyLight 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB102224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102224 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102224 Supplier Novus Biologicals Supplier No. NBP297478V

Rabbit Polyclonal Antibody

Mxi1 Polyclonal specifically detects Mxi1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Mxi1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias BHLHC11, Class C basic helix-loop-helix protein 11, MAX dimerization protein 2, MAX interacting protein 1, MAX interactor 1bHLHc11MAD2, max-interacting protein 1, Max-related transcription factor, MGC43220, MXD2, MXI
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Mxi1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4601
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.