Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MXRA8 Armenian Hamster anti-Human, Mouse, Clone: 9G2.D6, MilliporeSigma™
Armenian Hamster Monoclonal Antibody
$182.31 - $468.00
Specifications
Antigen | MXRA8 |
---|---|
Clone | 9G2.D6 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Inhibition Assays, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF227525
|
MilliporeSigma
MABF227525UL |
25 μL |
Each for $182.31
|
|
|||||
MABF2275100
|
MilliporeSigma
MABF2275100UL |
100 μL |
Each for $468.00
|
|
|||||
Description
MXRA8 Monoclonal antibody specifically detects MXRA8 in Human, Mouse samples. It is validated for Flow Cytometry, Inhibition, Western Blotting.Specifications
MXRA8 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified Armenian Hamster monoclonal antibody IgG1 in PBS without azide. | |
IgG1 κ | |
Protein G purified |
9G2.D6 | |
Flow Cytometry, Inhibition Assays, Western Blot | |
Unconjugated | |
Armenian Hamster | |
Human, Mouse | |
Fc-fusion recombinant fragment corresponding to aa 22-336 of extracellular domain of mouse MXRA8 protein. | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title