Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYBBP1A Antibody (3A1), Novus Biologicals™
SDP

Catalog No. NB298728 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298728 50 μL
NB298729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298728 Supplier Novus Biologicals Supplier No. NBP32612450UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MYBBP1A Monoclonal antibody specifically detects MYBBP1A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen MYBBP1A
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 3A1
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias MYB binding protein (P160) 1a, myb-binding protein 1A, P160FLJ37886, p53-activated protein-2, PAP2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human MYBBP1A [UniProt Q9BQG0]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10514
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.