Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYBBP1A Antibody, Novus Biologicals™
SDP

Catalog No. NB10061050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061050 Supplier Novus Biologicals Supplier No. NB10061050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

MYBBP1A Polyclonal specifically detects MYBBP1A in Human, Mouse samples. It is validated for CoIP, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, RIP-ChIP, Western Blot.

Specifications

Antigen MYBBP1A
Applications Co-immunoprecipitation (Co-IP), Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, RIP Assay, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:500 to 1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:500 to 1:2000, Proximity Ligation Assay
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_055335.1
Gene Alias MYB binding protein (P160) 1a, myb-binding protein 1A, P160FLJ37886, p53-activated protein-2, PAP2
Gene Symbols MYBBP1A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human MYB binding protein (P160) 1a using the numbering given in entry NP_055335.1 (GeneID 10514).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10514
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Chimpanzee.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.