Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYBPC1 Antibody (83B24), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309625 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309625 Supplier Novus Biologicals Supplier No. NBP250295MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYBPC1 Monoclonal antibody specifically detects MYBPC1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MYBPC1
Applications Western Blot
Classification Monoclonal
Clone 83B24
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type
Host Species Mouse
Immunogen Crude preparation of human skeletal myosin
Purification Method Ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 4604
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.