Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc Epitope Tag Antibody (9E10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB014138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014138 Supplier Novus Biologicals Supplier No. NBP281020AF350
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Myc Epitope Tag Monoclonal antibody specifically detects Myc Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen Myc Epitope Tag
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 90000000000
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias BHLHE39, Class E basic helix-loop-helix protein 39 (bHLHe39), Myc proto-oncogene protein, Proto-oncogene c-Myc, Transcription factor p64
Host Species Rat
Immunogen A synthetic peptide corresponding to aa 408-439 from C-terminus of human myc.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Epitope sequence, EQKLISEEDL, located in aa 410-419 of human c-myc protein.
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.