Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYCBP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10097841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10097841 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10097841 Supplier Novus Biologicals Supplier No. NB10097841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYCBP2 Polyclonal specifically detects MYCBP2 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).

Specifications

Antigen MYCBP2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation (Negative)
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O75592
Gene Alias DKFZp686M08244, EC 6.3.2.-, FLJ10106, FLJ21646, KIAA0916FLJ13826, MYC binding protein 2, Myc-binding protein 2, Pam/highwire/rpm-1 protein, PAMFLJ21597, probable E3 ubiquitin-protein ligase MYCBP2, Protein associated with Myc
Gene Symbols MYCBP2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2425 and 2475 of human protein associated with Myc using the numbering given in entry NP_055872.3 (GeneID 23077).
Molecular Weight of Antigen 510 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23077
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.