Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium avium Antibody (2A4), Biotin, Novus Biologicals™
SDP

Catalog No. NB10064559B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064559B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064559B Supplier Novus Biologicals Supplier No. NB10064559B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mycobacterium avium Monoclonal specifically detects Mycobacterium avium in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Mycobacterium avium
Applications ELISA
Classification Monoclonal
Clone 2A4
Conjugate Biotin
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias m avium, M. avium
Host Species Mouse
Immunogen Mycobacterium Avium cell extract.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for Mycobacterium avium. It does not react with Mycobacterium tuberculosis, Mycobacterium phlei, Corynebacterium bovis or Mycoplasma bovis.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.