Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium tuberculosis Antibody (BGN/1209/3875), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1062769A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062769A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062769A30 Supplier Novus Biologicals Supplier No. NB10062769AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mycobacterium tuberculosis Monoclonal specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Mycobacterium tuberculosis
Applications ELISA, Western Blot
Classification Monoclonal
Clone BGN/1209/3875
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias M. tuberculosis
Host Species Mouse
Immunogen Recombinant 38 kDa antigen from Mycobacterium tuberculosis.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This product recognises the 38 kDa antigen from Mycobacterium tuberculosis, a peptide believed to be involved in the regulation of proteins necessary for virulence.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.