Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium tuberculosis Antibody (BGN/1209/3875) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017509 0.025 mg
NB10062769 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017509 Supplier Novus Biologicals Supplier No. NB100627690.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mycobacterium tuberculosis Monoclonal specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Mycobacterium tuberculosis
Applications ELISA, Western Blot
Classification Monoclonal
Clone BGN/1209/3875
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Gene Alias M. tuberculosis
Host Species Mouse
Immunogen Recombinant 38 kDa antigen from Mycobacterium tuberculosis.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product recognises the 38 kDa antigen from Mycobacterium tuberculosis, a peptide believed to be involved in the regulation of proteins necessary for virulence.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.