Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium Tuberculosis CFP10 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB210247 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB210247 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB210247 Supplier Novus Biologicals Supplier No. NB11058737AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mycobacterium Tuberculosis CFP10 Polyclonal antibody specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for Western Blot

Specifications

Antigen Mycobacterium Tuberculosis CFP10
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias 10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10
Host Species Rabbit
Immunogen CFP10 (Rv3874) of Mycobacterium tuberculosis
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.