Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium Tuberculosis ESAT6 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB185986 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185986 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185986 Supplier Novus Biologicals Supplier No. NBP309114JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mycobacterium Tuberculosis ESAT6 Polyclonal antibody specifically detects Mycobacterium Tuberculosis ESAT6 in Bacteria samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Mycobacterium Tuberculosis ESAT6
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 6 kDa early secretory antigenic target, 6 kDa early secretory antigenic target ESXA, ESAT 6, esaT6, ESXA
Host Species Rabbit
Immunogen EsaT-6 (Rv3875) of Mycobacterium tuberculosis
Purification Method Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.