Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium Tuberculosis L-Alanine Dehydrogenase Antibody (1C3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP105061 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105061 Supplier Novus Biologicals Supplier No. NBP105061

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Mycobacterium Tuberculosis L-Alanine Dehydrogenase Monoclonal specifically detects Mycobacterium Tuberculosis L-Alanine Dehydrogenase in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Mycobacterium Tuberculosis L-Alanine Dehydrogenase
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Gene Alias 40 kDa antigen, Alanine dehydrogenase, EC 1.4.1.1, MT2850, MTV002.45, Rv2780, TB43
Host Species Mouse
Immunogen Culture filtrat from Mycobacterium tuberculosis H37Rv
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for a 40 kDa band from short term culture of Mycobacterium tuberculosis in Western blotting after SDS-PAGE. The 40 kDa band was identified as the protein L-alanine dehydrogenase (Ald). Interspecies analyses with HBT 10 showed reacts with M. tuberculosis and M. marinum but not M. bovis BCG.
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.