Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Antibody (1C3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | Mycobacterium Tuberculosis L-Alanine Dehydrogenase |
---|---|
Clone | 1C3 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105061
![]() |
Novus Biologicals
NBP105061 |
100 μg | N/A | N/A | N/A | ||||
Description
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Monoclonal specifically detects Mycobacterium Tuberculosis L-Alanine Dehydrogenase in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Mycobacterium Tuberculosis L-Alanine Dehydrogenase | |
1.0 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
Culture filtrat from Mycobacterium tuberculosis H37Rv | |
Primary | |
Store at 4C. Do not freeze. |
1C3 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
40 kDa antigen, Alanine dehydrogenase, EC 1.4.1.1, MT2850, MTV002.45, Rv2780, TB43 | |
IgG1 κ | |
Protein A or G purified | |
Specific for a 40 kDa band from short term culture of Mycobacterium tuberculosis in Western blotting after SDS-PAGE. The 40 kDa band was identified as the protein L-alanine dehydrogenase (Ald). Interspecies analyses with HBT 10 showed reacts with M. tuberculosis and M. marinum but not M. bovis BCG. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title