Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycoplasma pneumoniae Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10065504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065504 0.5 mL
NB017725 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10065504 Supplier Novus Biologicals Supplier No. NB10065504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mycoplasma pneumoniae Polyclonal specifically detects Mycoplasma pneumoniae in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Mycoplasma pneumoniae
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Phosphate buffered saline with 0.1% Sodium Azide
Gene Alias bacterial pneumonia, M. pneumoniae
Host Species Rabbit
Immunogen Purified Mycoplasma pneumoniae
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Rabbit anti Mycoplasma pneumoniae recognizes all antigens of Mycoplasma pneumoniae, a bacterium distinguished phenotypically from other bacteria by the lack of a peptidoglycan cell wall. Mycoplasma pneumoniae is a common causative agent of atypical pneumonia in young people. Central nervous system (CNS) manifestations are the most frequent extrapulmonary complications and include encephalitis, aseptic meningitis, cerebellar ataxia, and myelitis.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.