Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyD88 Antibody (316628), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB363735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363735 Supplier Novus Biologicals Supplier No. FAB29281W
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MyD88 Monoclonal specifically detects MyD88 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen MyD88
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 316628
Conjugate DyLight 650
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Host Species Rat
Immunogen E. coli-derived recombinant human MyD88, Met1-Pro296, Accession # Q99836
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4615
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.