Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyD88 Antibody (4D6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP227369W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227369W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227369W Supplier Novus Biologicals Supplier No. NBP227369AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MyD88 Monoclonal specifically detects MyD88 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).

Specifications

Antigen MyD88
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4D6
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular)
Gene Accession No. Q99836
Gene Alias MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Gene Symbols MYD88
Host Species Mouse
Immunogen A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4615
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human (isoform 1: 317 amino acids , isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa), Gorilla, Chimp, Rhesus Monkey and Orangutan, 82% homologous in Horse, 73% homologous in Dog, 68% homologous in Cat and Mouse (isoform CRA_a: 314 amino acids, CRA_b: 177 amino acids), and 63% homologous in Rat.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.