Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyD88 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB207919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB207919 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB207919 Supplier Novus Biologicals Supplier No. NBP176465H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MyD88 Polyclonal antibody specifically detects MyD88 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, KnockDown

Specifications

Antigen MyD88
Applications Western Blot, ELISA, KnockDown
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Host Species Rabbit
Immunogen Antibody (2127) was raised against a peptide corresponding to 17 amino acids near carboxy terminus of human MYD88 isoform 1. The immunogen is located within the last 50 amino acids of MYD88. Amino Acid Squence: CTKSWFWTRLAKALSLP
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4615
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.