Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MyD88 Inhibitor Peptide Set
SDP

Applications: Flow Cytometry, ELISA, Immunohistochemistry, In vitro assay, In vivo assay, Functional (Inhibition), Binding Inhibition, Block/Neutralize

$734.00 - $1452.00

Specifications

Components 2 x 1mg: MyD88 Homodimerization Inhibitor peptide (lyophilized) DRQIKIWFQNRRMKWKKRDVLPGT (MyD88 homodimerization sequence: RDVLPGT). Molecular weight: 3100
2 x 1mg: Antennapedia Control peptide (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
5mg size will contain 5mg of each peptide (MyD88 Homodimerization Inhibitor peptide and Antennapedia Control peptide)
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Product Type MyD88 Inhibitor Peptide Set
Molecular Weight (g/mol) 3100
Inhibitors MyD88
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP229328
SDP
View Documents
Novus Biologicals™
NBP229328
2 mg
Each for $734.00
Only null left
Add to Cart
 
NBP2293285
SDP
View Documents
Novus Biologicals™
NBP2293285MG
5 mg
Each for $1,452.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

2 x 1mg: MyD88 Homodimerization Inhibitor peptide (lyophilized) DRQIKIWFQNRRMKWKKRDVLPGT (MyD88 homodimerization sequence: RDVLPGT). Molecular weight: 3100
2 x 1mg: Antennapedia Control peptide (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
5mg size will contain 5mg of each peptide (MyD88 Homodimerization Inhibitor peptide and Antennapedia Control peptide)
MyD88 Inhibitor Peptide Set
MyD88
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
3100
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.