Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyD88 Inhibitor Peptide Set, Novus Biologicals™
SDP

Catalog No. NBP2293281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2293281 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2293281 Supplier Novus Biologicals Supplier No. NBP2293281MG
Add to Cart
Add to Cart

Antibody

MyD88 specifically detects MyD88 in Human, Mouse, Rat, Bovine, Xenopus, Yeast, Zebrafish samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, In vitro assay, In vivo assay, Functional (Inhibition), Binding Inhibition, Block/Neutralize.

Specifications

Applications Flow Cytometry, ELISA, Immunohistochemistry, In vitro Assay, In vivo Assay, Functional Inhibition Assay, Binding Inhibition Assay, Blocking Assay, Neutralization
Host Species Human, Mouse, Rat, Xenopus, Zebrafish
Quantity 1 mg
Primary or Secondary Primary
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Product Type Reactivity: Hu, Mu, Rt, Xp, Ze
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.