Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin PLP Antibody, Novus Biologicals™
SDP

Catalog No. NB1001608 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001608 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001608 Supplier Novus Biologicals Supplier No. NB1001608
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Myelin PLP Polyclonal specifically detects Myelin PLP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Myelin PLP
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:5000, Immunohistochemistry 1:2000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen
Formulation 10mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide
Gene Accession No. NP_000524, NP_955772, P60202
Gene Alias HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2
Gene Symbols PLP1
Host Species Chicken
Immunogen Chickens were immunized with a synthetic peptide/keyhole limpet hemocyanin (KLH) conjugate. The peptide corresponded to a region of the PLP gene product shared between the human (NP_000524, PLP1 isoform 1 and PLP1 isoform 2 (NP_955772)] and mouse (P60202). After
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5354
Test Specificity Proteolipid Protein
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.