Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin PLP Antibody (PLP1/4259), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB386200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386200 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386200 Supplier Novus Biologicals Supplier No. NBP308534AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myelin PLP Monoclonal antibody specifically detects Myelin PLP in Human, Mouse, Rat, Bovine, Reptile samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Myelin PLP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PLP1/4259
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2
Host Species Mouse
Immunogen Synthetic peptide GRGTKF that recognizes the C terminal region (aa272-277) of Myelin PLP protein. (Uniprot: P60201)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5354
Target Species Human, Mouse, Rat, Bovine, Reptile
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.