Learn More
Myeloid Cell Marker Antibody (BM-1), Allophycocyanin/Cy7, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Myeloid Cell Marker |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BM-1 |
| Conjugate | APC-Cyanine7 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein |
| Host Species | Mouse |
| Immunogen | Human peripheral blood mononuclear cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.