Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloid leukemia factor 1 Antibody (OTI3F11), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB287755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287755 Supplier Novus Biologicals Supplier No. NBP272849APC

Mouse Monoclonal Antibody

Myeloid leukemia factor 1 Monoclonal antibody specifically detects Myeloid leukemia factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Myeloid leukemia factor 1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3F11
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Myelodysplasia-myeloid leukemia factor 1, myeloid leukemia factor 1
Host Species Mouse
Immunogen Full length human recombinant protein of human MLF1(NP_071888) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4291
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.