Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase/MPO Antibody (2C7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1064803A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064803A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064803A32 Supplier Novus Biologicals Supplier No. NB10064803AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Myeloperoxidase/MPO
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2C7
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Gene Symbols MPO
Host Species Mouse
Immunogen Human Myeloperoxidase
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4353.0
Test Specificity NB100-64803 recognizes human myeloperoxidase (MPO). MPO is an important component of azurophilic granules in neutrophils, being involved in microbicidal processes. The protein is a multimer of 2 heavy chains (55kD) and two light chains (15kD), the heavy chains being linked by a disulphide bond. NB100-64803 recognizes native MPO in Western blots, and the heavy chain following boiling of the sample. The antibody also recognizes recombinant MPO in Western blots and weakly in ELISA. The antibody may be of value in the study of myeloid cells and myeloid leukaemias by flow cytometry following cell permeablisation.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.