Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase/MPO Antibody (2C7), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $353.50

Specifications

Antigen Myeloperoxidase/MPO
Clone 2C7
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:1000-1:5000
Applications Immunohistochemistry (Frozen), Flow Cytometry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017646
SDP
View Documents
Novus Biologicals
NB100648030.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10064803
SDP
View Documents
Novus Biologicals
NB10064803
0.1 mg
Each for $353.50
Only null left
Add to Cart
 
Description

Description

Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Myeloperoxidase/MPO
1 mg/mL
Immunohistochemistry (Frozen), Flow Cytometry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Unconjugated
Mouse
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
P05164
4353
Human Myeloperoxidase
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
2C7
Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:1000-1:5000
Monoclonal
Purified
RUO
Human
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
MPO
IgG1
Protein G purified
NB100-64803 recognizes human myeloperoxidase (MPO). MPO is an important component of azurophilic granules in neutrophils, being involved in microbicidal processes. The protein is a multimer of 2 heavy chains (55kD) and two light chains (15kD), the heavy chains being linked by a disulphide bond. NB100-64803 recognizes native MPO in Western blots, and the heavy chain following boiling of the sample. The antibody also recognizes recombinant MPO in Western blots and weakly in ELISA. The antibody may be of value in the study of myeloid cells and myeloid leukaemias by flow cytometry following cell permeablisation.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.