Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase/MPO Antibody (MPO421-8B2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP241406AJ Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241406AJ 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP241406AJ Supplier Novus Biologicals Supplier No. NBP241406APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen Myeloperoxidase/MPO
Applications Flow Cytometry
Classification Monoclonal
Clone MPO421-8B2
Conjugate APC
Dilution Flow Cytometry, Flow (Intracellular)
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Gene Symbols MPO
Host Species Mouse
Immunogen Human myeloperoxidase
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4353
Test Specificity The mouse monoclonal antibody MPO421-8B2 recognizes human myeloperoxidase, a heme protein present in myeloblasts, neutrophils and monocytes. It is a marker of acute myelogenous leukemias and acute lymphoblastic leukemias.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.