Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP106640 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106640 0.1 mL
NBP106640SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP106640 Supplier Novus Biologicals Supplier No. NBP106640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 23772023), Immunohistochemistry reported in scientific literature (PMID 35177643), Immunocytochemistry/Immunofluorescence 1:100
Formulation PBS (pH 7.4) with 0.1% Sodium Azide
Gene Alias EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B
Gene Symbols KDM6B
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse JMJD3 protein (within residues 600-650). [Swiss-Prot# Q5NCY0]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 23135
Test Specificity Human and mouse. 100% sequence identity with rat protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.