Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 7 publications

$244.50 - $560.50

Specifications

Antigen Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 23772023), Immunohistochemistry reported in scientific literature (PMID 35177643), Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP106640SS
SDP
View Documents
Novus Biologicals
NBP106640SS
0.025 mL
Each for $244.50
Only null left
Add to Cart
 
NBP106640
SDP
View Documents
Novus Biologicals
NBP106640
0.1 mL
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3
Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 23772023), Immunohistochemistry reported in scientific literature (PMID 35177643), Immunocytochemistry/Immunofluorescence 1:100
Polyclonal
Rabbit
Cancer, Chromatin Research, Transcription Factors and Regulators
PBS (pH 7.4) with 0.1% Sodium Azide
23135
Synthetic peptide made to an internal portion of the mouse JMJD3 protein (within residues 600-650). [Swiss-Prot# Q5NCY0]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse
EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B
KDM6B
IgG
Affinity Purified
Human and mouse. 100% sequence identity with rat protein.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.